Learn More
Invitrogen™ PSEN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA594969
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Brain Tissue, MCF-7 Cell, HELA Cell, SMMC Cell, CEM Cell. IHC: Human Mammary Cancer Tissue, Rat Cardiac Muscle Tissue, Rat Kidney Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1 or PSEN2) or the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an enzyme that cleaves APP. Also, it is thought that the presenilins are involved in the cleavage of the Notch receptor such that, they either directly regulate gamma-secretase activity, or themselves act are protease enzymes. Two alternatively spliced transcript variants encoding different isoforms of PSEN2 have been identified.
Specifications
| PSEN2 | |
| Polyclonal | |
| Unconjugated | |
| PSEN2 | |
| AD3L; AD3LP; AD4; Ad4h; AD5; Alg3; ALG-3; Alzheimer disease 4; CMD1V; E5-1; presenilin 2; presenilin-2; Presenilin-2 CTF subunit; Presenilin-2 NTF subunit; PS2; Ps-2; PSEN 2; PSEN2; Psnl2; STM2; STM-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19165, 5664, 81751 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| O88777, P49810, Q61144 | |
| PSEN2 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human Presenilin 2 (39-51aa EDGENTAQWRSQE). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.