missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pseudomonas Aeruginosa Antibody (1200/472), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08420C
This item is not returnable.
View return policy
Description
Pseudomonas Aeruginosa Monoclonal antibody specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA
Specifications
| Pseudomonas Aeruginosa | |
| Monoclonal | |
| DyLight 650 | |
| Pseudomonas aeruginosa 9 | |
| Pseudomonas aeruginosa serotype 6C | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| 1200/472 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction