missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pseudomonas Aeruginosa Antibody (95/159), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64562AF700
This item is not returnable.
View return policy
Description
Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Pseudomonas Aeruginosa | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Specific for Pseudomonas aeruginosa, serotype 9. | |
| Bacteria | |
| Purified |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 95/159 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Pseudomonas aeruginosa 9 | |
| Pseudomonas aeruginosa, serotype 9 (NCTC 11448, ATCC 33356). | |
| 0.1 mL | |
| Primary | |
| Protein A purified | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction