missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pseudomonas aeruginosa Monoclonal Antibody (B11)
GREENER_CHOICE

Product Code. 11504663
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11504663 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11504663 Supplier Invitrogen™ Supplier No. MA183430

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse anti Pseudomonas aeruginosaantibody, clone B11 recognizes Pseudomonas aeruginosa a gram-negative, opportunistic pathogen, commonly found in soil and water.

Pseudomonas Aeruginosa is a gram-negative, opportunistic pathogen commonly found in soil and water. In humans, P. aeruginosa can infect the urinary tract, respiratory and gastrointestinal system, soft tissues, bones and joints leading to severe systemic infections of immunosuppressed hospitalized patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pseudomonas aeruginosa
Applications ELISA
Classification Monoclonal
Clone B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.2
Gene Alias p aeruginosa; P. aeruginosa
Host Species Mouse
Immunogen Purified outer membrane protein of Pseudomonas aeruginosa.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.