Learn More
Invitrogen™ Pseudomonas Polyclonal Antibody, Biotin

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173119
Description
The PA1-73119 antibody reacts with Pseudomonas species. PA1-73119 has been successfully used in immunofluorescence applications. Possible applications for this product include avidin and streptavidin amplification systems for immunohistochemistry, ELISA, fluorescence microscopy, and immunoblotting. The PA1-73119 antibody was raised against whole cells of Pseudomonas aeruginosa, ATCC #27853. Recommended short term storage of less than 6 months at 2-8ºC. For extended storage, aliquot and freeze.
Although pathogenic bacteria constitute only a small portion of bacterial species, infectious diseases caused by pathogenic bacteria affect millions of people worldwide. Diarrheal diseases, many of which are bacterial, are the second leading cause of death worldwide, claiming an estimated 5 million people annually. Furthermore, an estimated 3 million people die of tuberculosis annually, 500,000 people die of pertussis, and 25,000 die of typhoid.
Specifications
| Pseudomonas | |
| Polyclonal | |
| Biotin | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Whole cells of Pseudomonas aeruginosa, ATCC #27853. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.