missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pseudomonas Polyclonal Antibody, FITC

Product Code. 11538491
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11538491 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11538491 Supplier Invitrogen™ Supplier No. PA173117

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The PA1-73117 antibody reacts with soluble and structural antigens from whole bacterium Pseudomonas species. This antiserum is not absorbed and may cross-react with related microorganisms PA1-73117 has been successfully used in direct FA staining of target antigens in a permissive tissue culture system. Acetone fixation of the antigen source is recommended prior to staining. The PA1-73117 antibody was raised against whole cells of Pseudomonas aeruginosa, ATCC #27853. Short term storage of less than 6 months, store at 4°C, in the dark. For long term storage, store at -20°C.

Although pathogenic bacteria constitute only a small portion of bacterial species, infectious diseases caused by pathogenic bacteria affect millions of people worldwide. Diarrheal diseases, many of which are bacterial, are the second leading cause of death worldwide, claiming an estimated 5 million people annually. Furthermore, an estimated 3 million people die of tuberculosis annually, 500,000 people die of pertussis, and 25,000 die of typhoid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pseudomonas
Applications Immunocytochemistry
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Host Species Rabbit
Immunogen Whole cells of Pseudomonas aeruginosa, ATCC #27853.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.