Learn More
PSF1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32156G
This item is not returnable.
View return policy
Description
PSF1 Polyclonal antibody specifically detects PSF1 in Human samples. It is validated for Western Blot
Specifications
| PSF1 | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Store at 4°C in the dark. |
| Western Blot | |
| Western Blot | |
| GINS complex subunit 1, GINS complex subunit 1 (Psf1 homolog), KIAA0186PSF1DNA replication complex GINS protein PSF1, partner of sld five-1 |