missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PSF2 Polyclonal antibody specifically detects PSF2 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | PSF2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | DNA replication complex GINS protein PSF2, GINS complex subunit 2, GINS complex subunit 2 (Psf2 homolog), HSPC037, PSF2Pfs2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-185 of human PSF2 (NP_057179.1).,, Sequence:, MDAAEVEFLAEKELVTIIPNFSLDKIYLIGGDLGPFNPGLPVEVPLWLAINLKQRQKCRLLPPEWMDVEKLEKMRDHERKEETFTPMPSPYYMELTKLLLNHASDNIPKADEIRTLVKDMWDTRIAKLRVSADSFVRQQEAHAKLDNLTLMEINTSGTFLTQALNHMYKLRTNLQPLESTQSQDF |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?