missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSG1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | PSG1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687302
|
Novus Biologicals
NBP2-95140-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644142
|
Novus Biologicals
NBP2-95140-0.1ml |
0.1 mL |
€ 470.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSG1 Polyclonal antibody specifically detects PSG1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| PSG1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| CD Markers, Immunology | |
| PBS (pH 7.3), 50% glycerol | |
| 5669 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| B1G1CD66 antigen-like family member F, CD66f, CD66f antigen, DHFRP2, Fetal liver non-specific cross-reactive antigen 1/2, FLJ90598, FLJ90654, FL-NCA-1/2, PBG1, pregnancy specific beta-1-glycoprotein 1, pregnancy-specific B-1 glycoprotein, Pregnancy-specific beta-1 glycoprotein C/D, pregnancy-specific beta-1-glycoprotein 1, Pregnancy-specific glycoprotein 1, PS-beta-C/D, PS-beta-G-1, PSBG-1, PSBG1SP1, PSGGAPSG95, PSGIIA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 180-419 of human PSG1 (NP_001171754.1). WWMNGQSLPMTHSLKLSETNRTLFLLGVTKYTAGPYECEIRNPVSASRSDPVTLNLLPKLPKPYITINNLNPRENKDVLNFTCEPKSENYTYIWWLNGQSLPVSPRVKRPIENRILILPSVTRNETGPYQCEIRDRYGGIRSDPVTLNVLYGPDLPRIYPSFTYYRSGEVLYLSCSADSNPPAQYSWTINEKFQLPGQKLFIRHITTKHSGLYVCSVRNSATGKESSKSMTVEVSDWTVP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title