missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Psg16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586002
This item is not returnable.
View return policy
Description
Psg16 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Psg16 | |
| Polyclonal | |
| Unconjugated | |
| Psg16 | |
| bCEA; carcinoembryonic antigen 11; Cea11; pregnancy specific glycoprotein 16; PSG; Psg16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26436 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| 0 | |
| Psg16 | |
| Synthetic peptide within Human Psg16 aa 346-395. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction