missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSG2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15827603
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15827603 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15827603 Supplier Invitrogen™ Supplier No. PA569190

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

The human pregnancy-specific glycoproteins (PSGs) are a family of proteins that are synthesized in large amounts by placental trophoblasts and released into the maternal circulation during pregnancy. Molecular cloning and analysis of several PSG genes has indicated that the PSGs form a subgroup of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily of genes. Members of the CEA family consist of a single N domain, with structural similarity to the immunoglobulin variable domains, followed by a variable number of immunoglobulin constant-like A and/or B domains. Most PSGs have an arg-gly-asp (RGD) motif, which has been shown to function as an adhesion recognition signal for several integrins, in the N-terminal domain (summary by Teglund et al., 1994 [PubMed 7851896]). For additional general information about the PSG gene family, see PSG1 (MIM 176390).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PSG2
Gene Accession No. P11465
Gene Alias carcinoembryonic antigen SG8; CEA; pregnancy specific beta-1-glycoprotein 2; pregnancy-specific beta-1 glycoprotein E; Pregnancy-specific beta-1-glycoprotein 2; pregnancy-specific beta-1-glycoprotein 7; pregnancy-specific beta-1-glycoprotein-2; pregnancy-specific glycoprotein 2; PS-beta-E; PS-beta-G-2; PSBG2; PSBG-2; PSG1; PSG2
Gene Symbols PSG2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human PSG2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5670
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.