missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSG5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17186813
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17186813 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17186813 Supplier Invitrogen™ Supplier No. PA5107008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSG5 Polyclonal Antibody for Western Blot, IHC, IHC (P)

The pregnancy-specific beta 1 glycoprotein (PSG) is a group of heterogeneous proteins produced in large amounts by the human syncytiotrophoblast. They belong to the carcinoembryonic antigen (CEA) family. The function of PSG5 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene PSG5
Gene Accession No. Q15238
Gene Alias Fetal liver non-specific cross-reactive antigen 3; FL-NCA-3; pregnancy specific beta-1-glycoprotein 5; pregnancy-specific beta 1 glycoprotein; pregnancy-specific beta-1 glycoprotein; pregnancy-specific beta-1-glycoprotein 5; Pregnancy-specific beta-1-glycoprotein-5; pregnancy-specific glycoprotein 5; PS-beta-G-5; PSBG-5; PSG; PSG5
Gene Symbols PSG5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PSG5 (aa 287-335).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5673
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.