missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSGL-1/CD162 Antibody (108), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90249G
This item is not returnable.
View return policy
Description
PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| PSGL-1/CD162 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 108 | |
| Western Blot, ELISA | |
| CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PSGL-1/CD162 (Q14242; Met1-Gly295). | |
| 0.1 mL | |
| Cholesterol Metabolism, Immunology, Lipid and Metabolism | |
| 6404 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction