missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSGL-1/CD162 Antibody (PSGL1/8192R) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20932JF669
This item is not returnable.
View return policy
Description
PSGL-1/CD162 Monoclonal antibody specifically detects PSGL-1/CD162 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PSGL-1/CD162 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA | |
| Recombinant full-length human PSGL-1/CD162 protein | |
| 0.1 mL | |
| Cholesterol Metabolism, Immunology, Lipid and Metabolism | |
| 6404 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| PSGL1/8192R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction