Learn More
PSI-6206 13C,d3, MedChemExpress
Product Code. 16451489
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16451489
Brand: Medchem Express HY15236S
MedChemExpress PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.
PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.
Specifications
Chemical Name or Material PSI-6206 13C,d3
CAS 1256490-42-2
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C9 13CH10D3FN2O5
Color Light Yellow
Synonym RO-2433 13C,d3 GS-331007 13C,d3 Sofosbuvir metabolite GS-331007 13C,d3
SMILES OC[C@@H]1[C@H]([C@@]([13C]([2H])([2H])[2H])(F)[C@H](N2C(NC(C=C2)=O)=O)O1)O
Molecular Weight (g/mol) 264.23
Quantity 1 mg
Formula Weight 264.23
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only