Learn More
Invitrogen™ PSIP1 Recombinant Rabbit Monoclonal Antibody (24GB840)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557649
 View more versions of this product
Product Code. 30276280
  • € 204.00 / 20µL
Description
PSIP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Transcriptional coactivator involved in neuroepithelial stem cell differentiation and neurogenesis. Involved in particular in lens epithelial cell gene regulation and stress responses. May play an important role in lens epithelial to fiber cell terminal differentiation. May play a protective role during stress-induced apoptosis. Isoform 2 is a more general and stronger transcriptional coactivator. Isoform 2 may also act as an adapter to coordinate pre-mRNA splicing. Cellular cofactor for lentiviral integration.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PSIP1
Recombinant Monoclonal
Unconjugated
PSIP1
AA408851; AU015605; CLL-associated antigen KW-7; dense fine speckles 70; dense fine speckles 70 kDa protein; DFS 70; DFS70; LEDGF; Ledgfa; Ledgfb; Lens epithelium-derived growth factor; mLEDGF; p52; p75; PAIP; PC4 and SFRS1 interacting protein 1; PC4 and SFRS1 interacting protein 2; PC4 and SFRS1-interacting protein; PSIP1; Psip2; SBP75; supercoiled DNA binding protein 75; transcriptional coactivator p52/p75; Transcriptional coactivator p75/p52
Rabbit
Affinity chromatography
RUO
101739, 11168, 313323
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB840
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75475, Q812D1, Q99JF8
PSIP1
A synthesized peptide derived from human PSIP1 / LEDGF (aa 1-61).