missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSKH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575280
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
PSKH1 may be a SFC-associated serine kinase (splicing factor compartment-associated serine kinase) with a role in intranuclear SR protein (non-snRNP splicing factors containing a serine/arginine-rich domain) trafficking and pre-mRNA processing. PSKH1 is localized in the Brefeldin A-sensitive Golgi compartment, at centrosomes, in the nucleus with a somewhat speckle-like presence, membrane-associated to the endoplasmic reticulum (ER) and the plasma membrane (PM), and more diffusely in the cytoplasm.
Specifications
| PSKH1 | |
| Polyclonal | |
| Unconjugated | |
| Pskh1 | |
| AW539964; b2b1230Clo; E130013P03Rik; Mutant line 1230; protein serine kinase H1; PSKH1; PSK-H1; Serine/threonine-protein kinase H1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 244631, 364993, 5681 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P11801, Q91YA2 | |
| Pskh1 | |
| Synthetic peptide corresponding to amino acids 261-310 of Human PSKH1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction