missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSKH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578611
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. PSKH2 (protein serine kinase H2) is a 385 amino acid protein belonging to the CAMK Ser/Thr protein kinase family. Containing one protein kinase domain, PSKH2 is encoded by a gene located on human chromosome 8, which consists of nearly 146 million base pairs, houses more than 800 genes and is associated with a variety of diseases and malignancies. Schizophrenia, bipolar disorder, Trisomy 8, Pfeiffer syndrome, congenital hypothyroidism, Waardenburg syndrome and some leukemias and lymphomas are thought to occur as a result of defects in specific genes that map to chromosome 8.
Specifications
| PSKH2 | |
| Polyclonal | |
| Unconjugated | |
| PSKH2 | |
| protein serine kinase H2; PSKH2; PSK-H2; serine/threonine kinase PSKH2; serine/threonine-protein kinase H2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85481 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.26 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96QS6 | |
| PSKH2 | |
| Recombinant protein encompassing a sequence within the center region of human PSKH2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction