missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA/FOLH1/NAALADase I Antibody (FOLH1/2121), DyLight 680, Novus Biologicals™
Shop All Bio Techne ProductsDescription
PSMA/FOLH1/NAALADase I Monoclonal antibody specifically detects PSMA/FOLH1/NAALADase I in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | PSMA/FOLH1/NAALADase I |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | FOLH1/2121 |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase |
| Host Species | Mouse |
| Immunogen | Recombinant human PSMA/FOLH1/NAALADase I protein fragment (around aa 232-433) (exact sequence is proprietary) (Uniprot: Q04609) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?