missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA/FOLH1/NAALADase I Antibody (GCP-04), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-45057AF647
This item is not returnable.
View return policy
Description
PSMA/FOLH1/NAALADase I Monoclonal antibody specifically detects PSMA/FOLH1/NAALADase I in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| PSMA/FOLH1/NAALADase I | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Q04609 | |
| FOLH1 | |
| Recombinant fragment of human GCPII (amino acids 44-750) produced in S2 cells (NM_001014986.1). | |
| 0.1 mL | |
| Cancer, Prostate Cancer | |
| 2346 | |
| Human, Mouse, Rat, Pig, Canine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GCP-04 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody GCP-04 recognizes amino acids 100-104 of extracellular domain of denaturated glutamate carboxypeptidase II (PSMA, NAALADase, FOLH1), an approximately 95-110 kDa transmembrane glycoprotein. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction