missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA/FOLH1/NAALADase I Antibody (rosopatamab) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28155JF525
This item is not returnable.
View return policy
Description
PSMA/FOLH1/NAALADase I Monoclonal antibody specifically detects PSMA/FOLH1/NAALADase I in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PSMA/FOLH1/NAALADase I | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase | |
| FOLH1/ PSMA | |
| 0.1 mL | |
| Cancer, Prostate Cancer | |
| 2346 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| rosopatamab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction