missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSMA Monoclonal Antibody (GCP-05), Alexa Fluor™ 488
GREENER_CHOICE

Product Code. 15387424
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15387424 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15387424 Supplier Invitrogen™ Supplier No. MA518161

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes extracellular domain (preferentially in native form) of glutamate carboxypeptidase II (NAALADase, FOLH1, PSMA), an approximately 95-110 kDa transmembrane glycoprotein expressed mainly in tumor neovasculatures, nervous system and jejunum, which is an important prostate tumor marker.

This gene encodes a type II transmembrane glycoprotein belonging to the M28 peptidase family. The protein acts as a glutamate carboxypeptidase on different alternative substrates, including the nutrient folate and the neuropeptide N-acetyl-l-aspartyl-l-glutamate and is expressed in a number of tissues such as prostate, central and peripheral nervous system and kidney. A mutation in this gene may be associated with impaired intestinal absorption of dietary folates, resulting in low blood folate levels and consequent hyperhomocysteinemia. Expression of this protein in the brain may be involved in a number of pathological conditions associated with glutamate excitotoxicity. In the prostate the protein is up-regulated in cancerous cells and is used as an effective diagnostic and prognostic indicator of prostate cancer. This gene likely arose from a duplication event of a nearby chromosomal region. Alternative splicing gives rise to multiple transcript variants encoding several different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMA
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone GCP-05
Concentration 1.0 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 15mM sodium azide
Gene FOLH1
Gene Accession No. Q04609
Gene Alias Cell growth-inhibiting gene 27 protein; FGCP; folate hydrolase (prostate-specific membrane antigen) 1; folate hydrolase 1; FOLH; Folh1; Folylpoly-gamma-glutamate carboxypeptidase; GCP2; GCPII; GIG27; glutamate carboxylase II; glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; membrane glutamate carboxypeptidase; mGCP; mopsm; Naalad; NAALAD1; NAALAdase; NAALADase I; N-acetylated alpha-linked acidic dipeptidase; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; prostate specific membrane antigen; prostate specific membrane antigen variant F; prostate-specific membrane antigen; Prostate-specific membrane antigen homolog; PSM; PSMA; pteroylpoly-gamma-glutamate carboxypeptidase
Gene Symbols FOLH1
Host Species Mouse
Immunogen Amino acids 44-750 of human GCPII.
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2346
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.