Learn More
PSMA Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA813986S
Encompass_Preferred
 View more versions of this product
Product Code. 17518734
  • € 242.00 / 30µL
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
PSMA Monoclonal antibody specifically detects PSMA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
PSMA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q04609
Mouse
Protein A/G
RUO
2346
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin)
OTI1F5
Unconjugated
FOLH1
Cell growth-inhibiting gene 27 protein; FGCP; folate hydrolase (prostate-specific membrane antigen) 1; folate hydrolase 1; FOLH; Folh1; Folylpoly-gamma-glutamate carboxypeptidase; GCP2; GCPII; GIG27; glutamate carboxylase II; glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; membrane glutamate carboxypeptidase; mGCP; mopsm; Naalad; NAALAD1; NAALAdase; NAALADase I; N-acetylated alpha-linked acidic dipeptidase; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; prostate specific membrane antigen; prostate specific membrane antigen variant F; prostate-specific membrane antigen; Prostate-specific membrane antigen homolog; PSM; PSMA; pteroylpoly-gamma-glutamate carboxypeptidase
Full length human recombinant protein of human PSMA (NP_004467) produced in HEK293T cell.
30 μL