missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA2 Antibody (OTI3D9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73687CL1
This item is not returnable.
View return policy
Description
PSMA2 Monoclonal antibody specifically detects PSMA2 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| PSMA2 | |
| Monoclonal | |
| CoraFluor 1 | |
| HC3;MU;PMSA2;Proteasome subunit alpha type;Proteasome subunit alpha type-2;PSC2 | |
| Human recombinant protein fragment corresponding to amino acids 50-234 of human PSMA2(NP_002778) produced in E.coli. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 5683 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| OTI3D9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction