missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCC Mouse anti-Human, Clone: OTI5D10, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804517S
This item is not returnable.
View return policy
Description
DCC Monoclonal antibody specifically detects DCC in Human samples. It is validated for Western BlotSpecifications
| DCC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P43146 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1630 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI5D10 | |
| Unconjugated | |
| DCC | |
| C030036D22Rik; Colorectal cancer suppressor; colorectal tumor suppressor; CRC18; CRCR1; Dcc; DCC netrin 1 receptor; deleted in colorectal cancer protein; deleted in colorectal carcinoma; IGDCC1; Immunoglobulin superfamily DCC subclass member 1; immunoglobulin superfamily, DCC subclass, member 1; MRMV1; Netrin receptor DCC; NTN1R1; putative colorectal tumor suppressor; tumor suppressor protein DCC | |
| Full length human recombinant protein of human DCC produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction