missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSMA2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17421863 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17421863 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17421863 Supplier Invitrogen™ Supplier No. PA5118201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMA2 Polyclonal Antibody for Western Blot, IHC (P), IP

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMA2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene Psma2
Gene Accession No. P25787
Gene Alias HC3; Lmpc3; Macropain subunit C3; MU; multicatalytic endopeptidase complex subunit C3; OTTHUMP00000207364; OTTHUMP00000207366; PMSA2; proteasome (prosome, macropain) subunit, alpha type 2; proteasome (prosome, macropain) subunit, alpha type, 2; proteasome 20S subunit alpha 2; proteasome alpha 2 subunit; proteasome component C3; proteasome subunit alpha 2; proteasome subunit alpha type, 2; proteasome subunit alpha type-2; proteasome subunit HC3; PSC2; PSC3; PSMA 2; PSMA2; wu:faa49c06; wu:fb98h03; zgc:110710
Gene Symbols Psma2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human PSMA2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5683
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.