missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA4 Antibody (OTI4B8), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73690C
This item is not returnable.
View return policy
Description
PSMA4 Monoclonal specifically detects PSMA4 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PSMA4 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PSMA4 | |
| Full length recombinant protein of human PSMA4 (NP_002780) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Primate, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4B8 | |
| Western Blot, Immunohistochemistry | |
| HC9EC 3.4.25.1, HsT17706, Macropain subunit C9, MGC12467, MGC24813, Multicatalytic endopeptidase complex subunit C9, proteasome (prosome, macropain) subunit, alpha type, 4, Proteasome component C9, proteasome subunit alpha type-4, proteasome subunit HC9, Proteasome subunit L, PSC9MGC111191 | |
| Mouse | |
| Affinity Purified | |
| Proteases & Other Enzymes | |
| 5685.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction