missing translation for 'onlineSavingsMsg'
Learn More

PSMA6 Monoclonal Antibody (OTI4C9), TrueMAB™, OriGene

Product Code. p-200015636
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17587582 30 μL 30µL
17567582 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17587582 Supplier Origene Technologies Supplier No. TA800101S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSMA6 Monoclonal antibody specifically detects PSMA6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PSMA6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Uqcrc1
Gene Accession No. P31930
Gene Alias 27 kDa prosomal protein; EGK_18113; IOTA; Macropain iota chain; macropain subunit iota; MGC22756; MGC2333; MGC23846; multicatalytic endopeptidase complex iota chain; p27K; PROS 27; PROS27; PROS-27; prosomal P27K protein; proteasome (prosome, macropain) subunit, alpha type 6; proteasome (prosome, macropain) subunit, alpha type, 6; proteasome 20S subunit alpha 6; proteasome alpha 6 subunit; proteasome iota chain; proteasome subunit alpha 6; proteasome subunit alpha type-6; proteasome subunit iota; PSA6; PSMA 6; PSMA6; testicular secretory protein Li 44
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 46-305 of human UQCRC1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7384
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.