Learn More
PSMA6 Mouse, anti-Human, CoraLite 488, Clone: 1D4B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67695-100UL
Description
PSMA6(Proteasome subunit alpha type-6) is also named as PROS27 and belongs to the peptidase T1A family.The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH.It also has an ATP-dependent proteolytic activity.
Specifications
| PSMA6 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |
| Immunofluorescence | |
| 1D4B2 | |
| CoraLite 488 | |
| 100 μL | |
| 5687 | |
| Liquid |