missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA7 Antibody (OTI3F11), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73693AF647
This item is not returnable.
View return policy
Description
PSMA7 Monoclonal specifically detects PSMA7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| PSMA7 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PSMA7 | |
| Full length recombinant protein of human PSMA7 (NP_002783) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| OTI3F11 | |
| Western Blot, Flow Cytometry | |
| C6, EC 3.4.25.1, HSPCMGC3755, proteasome (prosome, macropain) subunit, alpha type, 7, proteasome subunit alpha 4, proteasome subunit alpha type-7, Proteasome subunit RC6-1, Proteasome subunit XAPC7, RC6-1, XAPC7 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Core ESC Like Genes, Hypoxia, Stem Cell Markers | |
| 5688.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction