Learn More
PSMA7 Antibody (OTI3F11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-73693UV
Product Code. 18108006
  • € 751.00 / 0.10mL
Description
PSMA7 Monoclonal specifically detects PSMA7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PSMA7
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
PSMA7
Full length recombinant protein of human PSMA7 (NP_002783) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Flow Cytometry
OTI3F11
Western Blot, Flow Cytometry
C6, EC 3.4.25.1, HSPCMGC3755, proteasome (prosome, macropain) subunit, alpha type, 7, proteasome subunit alpha 4, proteasome subunit alpha type-7, Proteasome subunit RC6-1, Proteasome subunit XAPC7, RC6-1, XAPC7
Mouse
Affinity Purified
Cancer, Core ESC Like Genes, Hypoxia, Stem Cell Markers
5688.0
Store at 4°C in the dark.