Learn More
PSMA7 Monoclonal Antibody (OTI8B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810368
Encompass_Preferred
 View more versions of this product
Product Code. 17503404
  • € 540.87 / 100µL
Description
Component of the 20S core proteasome complex involved in the proteolytic degradation of most intracellular proteins. This complex plays numerous essential roles within the cell by associating with different regulatory particles. Associated with two 19S regulatory particles, forms the 26S proteasome and thus participates in the ATP-dependent degradation of ubiquitinated proteins. The 26S proteasome plays a key role in the maintece of protein homeostasis by removing misfolded or damaged proteins that could impair cellular functions, and by removing proteins whose functions are no longer required. Associated with the PA200 or PA28, the 20S proteasome mediates ubiquitin-independent protein degradation. This type of proteolysis is required in several pathways including spermatogenesis (20S-PA200 complex) or generation of a subset of MHC class I-presented antigenic peptides (20S-PA28 complex). Inhibits the transactivation function of HIF-1A under both normoxic and hypoxia-mimicking conditions. The interaction with EMAP2 increases the proteasome-mediated HIF-1A degradation under the hypoxic conditions. Plays a role in hepatitis C virus internal ribosome entry site-mediated translation. Mediates nuclear translocation of the androgen receptor (AR) and thereby enhances androgen-mediated transactivation.
Specifications
Specifications
PSMA7
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O14818
PSMA7
Full length human recombit protein of human PSMA7 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI8B11
Unconjugated
PSMA7
C6, HEL-S-276, HSPC, RC6-1, XAPC7
Mouse
Affinity Chromatography
RUO
5688
-20° C, Avoid Freeze/Thaw Cycles