missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA7 Monoclonal Antibody (OTI8B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810368S
This item is not returnable.
View return policy
Description
PSMA7 Monoclonal antibody specifically detects PSMA7 in Human samples. It is validated for Western BlotSpecifications
| PSMA7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q53HV7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23583 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8B11 | |
| Unconjugated | |
| SMUG1 | |
| C6; C6-I; epididymis secretory protein Li 276; HEL-S-276; HSPC; proteasome (prosome, macropain) subunit, alpha type 7; proteasome (prosome, macropain) subunit, alpha type, 7; proteasome subunit alpha 4; proteasome subunit alpha 7; proteasome subunit alpha type-7; Proteasome subunit RC6-1; proteasome subunit XAPC7; Psma7; RC6-1; RP5-1005F21.4; testicular tissue protein Li 151; XAPC7 | |
| Full length human recombinant protein of human SMUG1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction