missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB10 Mouse anti-Human, Porcine, Clone: 1G8C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66403-1-IG-150UL
This item is not returnable.
View return policy
Description
The 20S proteasome is a protease complex that is responsible for cytosolic protein degradation and generation of peptide ligands for major histocompatabilty complex (MHC) class I molecules, either in their final form or in the form of amino-terminally extended precursors. Upon IFN-gamma stimulation of cells, three constitutively expressed subunits of the 20S proteasome are replaced by inducible subunits LMP2 (low-molecular mass polypeptide 2), LMP7, and MECL-1 (multicatalytic endopeptidase complex-like-1, LMP10). LMP2, LMP7, and MECL-1 subunits form immunoproteasomes, which are associated with more efficient class I antigen processing and presentation. Independent assortment of LMP-2, LMP-7, and MECL-1 into different proteasome complexes can lead to 36 unique proteasome subsets, which may mediate differences in the cleavage specificities/cleavage motifs of proteins subject to constitutive- and immuno-proteasomes.Specifications
| PSMB10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P40306 | |
| PSMB10 | |
| PSMB10 Fusion Protein Ag8835 | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1G8C1 | |
| Unconjugated | |
| PSMB10 | |
| beta2i, LMP10, Low molecular mass protein 10, Macropain subunit MECl 1, MECL1, Proteasome MECl 1, Proteasome subunit beta 2i, PSMB10 | |
| Mouse | |
| Protein A | |
| RUO | |
| 5699, 733626 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction