missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSMB10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15774311 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15774311 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15774311 Supplier Invitrogen™ Supplier No. PA556403

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FRYQGHVGAS LIVGGVDLTG PQLYGVHPHG SYSRLPFTAL GSGQDAALAV LEDRFQPNMT LEAAQGLLVE AVT Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.

The 20S proteasome is a protease complex that is responsible for cytosolic protein degradation and generation of peptide ligands for major histocompatibility complex (MHC) class I molecules, either in their final form or in the form of amino-terminally extended precursors. Upon IFN-gamma stimulation of cells, three constitutively expressed subunits of the 20S proteasome are replaced by inducible subunits LMP2 (low-molecular mass polypeptide 2), LMP7, and MECL-1 (multicatalytic endopeptidase complex-like-1, LMP10). LMP2, LMP7, and MECL-1 subunits form immunoproteasomes, which are associated with more efficient class I antigen processing and presentation. Independent assortment of LMP-2, LMP-7, and MECL-1 into different proteasome complexes can lead to 36 unique proteasome subsets, which may mediate differences in the cleavage specificities/cleavage motifs of proteins subject to constitutive- and immuno-proteasomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSMB10
Gene Accession No. P40306
Gene Alias beta2i; LMP10; Low molecular mass protein 10; macropain; Macropain subunit MECl-1; MECL1; Mecl-1; MGC1665; multicatalytic endopeptidase complex subunit MECl-1; OTTHUMP00000174858; prosome; prosome Mecl1; proteasome (prosome, macropain) subunit, beta type 10; proteasome (prosome, macropain) subunit, beta type, 10; proteasome (prosomome, macropain) subunit, beta type 10; proteasome 20S subunit beta 10; proteasome beta 10 subunit; proteasome catalytic subunit 2i; Proteasome MECl 1; proteasome MECl-1; proteasome subunit 10; proteasome subunit beta 10; proteasome subunit beta 7i; proteasome subunit beta type-10; Proteasome subunit beta-2i; proteasome subunit LMP10; proteasome subunit MECL1; PSMB 10; Psmb10
Gene Symbols PSMB10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSMB10. Recombinant protein control fragment (Product #RP-102567).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5699
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.