missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144668
This item is not returnable.
View return policy
Description
Positive control: Hela cell lysate, K562 cell lysate, 293T cell lysate, Jurkat cell lysate, Hela, Human skin tissue, Human breast tissue. Predicted band size: 23 kDa Subcellular Location: Cytoplasm, Nucleus, Proteasome.
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. Pseudogenes have been identified on chromosomes 2 and 12.
Specifications
| PSMB3 | |
| Polyclonal | |
| Unconjugated | |
| PSMB3 | |
| AL033320; C10-II; HC10-II; MGC4147; proteasome (prosome, macropain) subunit, beta type 3; proteasome (prosome, macropain) subunit, beta type, 3; proteasome beta 3 subunit; proteasome chain 13; Proteasome component C10-II; proteasome subunit beta 3; proteasome subunit beta type-3; Proteasome theta chain; PSMB3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5691 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P49720 | |
| PSMB3 | |
| Recombinant full length protein within human PSMB3 aa 1-205/205. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction