missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB4 Monoclonal Antibody (OTI7B4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503481S
This item is not returnable.
View return policy
Description
PSMB4 Monoclonal antibody specifically detects PSMB4 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PSMB4 | |
| Monoclonal | |
| 0.42 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P28070 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5692 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI7B4 | |
| Unconjugated | |
| PSMB4 | |
| 26 kDa prosomal protein; HN3; HsBPROS26; HsN3; Lmp3; Low molecular mass protein 3; macropain beta chain; Multicatalytic endopeptidase complex beta chain; PROS26; PROS-26; Pros-27; proteasome (prosome, macropain) subunit, beta type 4; proteasome (prosome, macropain) subunit, beta type, 4; proteasome beta 4 subunit; proteasome beta chain; Proteasome chain 3; proteasome subunit beta 4; proteasome subunit beta type-4; proteasome subunit HsN3; proteasome subunit, beta type, 4; Psmb4; RN3; testis tissue sperm-binding protein Li 79P | |
| Full length human recombinant protein of human PSMB4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction