Learn More
Invitrogen™ PSMB5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1977
Description
PA1-977 detects proteasome 20S X from bovine, mouse and human samples. PA1-977 has been successfully used in Western blot and ICC/IF procedures. By Western blot, this antibody detects an ~23 kDa protein representing proteasome 20S X. The PA1-977 immunizing peptide corresponds to amino acid residues 243-259 from mouse proteasome 20S X. This sequence is 94% conserved in human and rat. PA1-977 immunizing peptide (Cat. # PEP-172) is available for use in neutralization and control procedures.
Cyclic nucleotide phosphodiesterases.
Specifications
| PSMB5 | |
| Polyclonal | |
| Unconjugated | |
| PSMB5 | |
| LMPX; Macropain epsilon chain; MB1; MGC118075; Multicatalytic endopeptidase complex epsilon chain; proteasome (prosome, macropain) subunit, beta type 5; proteasome (prosome, macropain) subunit, beta type, 5; proteasome 20S subunit beta 5; proteasome beta 5 subunit; proteasome beta type subunit 5; proteasome catalytic subunit 3; proteasome chain 6; Proteasome epsilon chain; proteasome subunit beta 5; proteasome subunit beta type-5; Proteasome subunit MB1; proteasome subunit X; proteasome subunit, beta type, 5; PSMB5; PSX large multifunctional protease X; testicular tissue protein Li 153; Unknown (protein for MGC:134464); X | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19173, 534640, 5693 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| O55234, P28074, Q32KL2 | |
| PSMB5 | |
| Synthetic Peptide: C W(243) I R V S S D N V A D L H D K Y S(259). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Bovine | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.