missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB7 Antibody (OTI1E1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73645F
This item is not returnable.
View return policy
Description
PSMB7 Monoclonal specifically detects PSMB7 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Immunohistochemistry.
Specifications
| PSMB7 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Monkey, Primate | |
| IgG1 |
| Immunohistochemistry | |
| OTI1E1 | |
| Immunohistochemistry | |
| PSMB7 | |
| Recombinant protein fragment corresponding to amino acids 58-277 of human Proteasome 20S beta 7 (NP_002790) produced in E.coli. | |
| 0.1 mL | |
| 5695.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction