missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 195.00
Specifications
| Antigen | PSMC1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18547251
|
Novus Biologicals
NB120-3317-0.025ml |
0.025 mL | |||||||
Description
PSMC1 Polyclonal specifically detects PSMC1 in Human, Yeast samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| PSMC1 | |
| Western Blot 1:1000, Immunoprecipitation 1:10 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
| 26S protease regulatory subunit 4, 26S proteasome AAA-ATPase subunit RPT2, MGC24583, P26S4, p56, proteasome (prosome, macropain) 26S subunit, ATPase, 1, proteasome 26S ATPase subunit 1, Proteasome 26S subunit ATPase 1, proteasome 26S subunit, ATPase, 1, S4MGC8541 | |
| PSMC1 | |
| IgG | |
| Ammonium sulfate precipitation | |
| Detects proteasome 19S subunit S4. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P62191 | |
| 5700 | |
| Yeast proteasome 19S subunit S4. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title