missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC3 Antibody (OTI9F3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73696AF350
This item is not returnable.
View return policy
Description
PSMC3 Monoclonal specifically detects PSMC3 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| PSMC3 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PSMC3 | |
| Full length recombinant protein of human PSMC3 (NP_002795) produced in HEK293T cells. | |
| 0.1 mL | |
| 5702.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI9F3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| human immunodeficiency virus tat transactivator binding protein-1, MGC8487, proteasome (prosome, macropain) 26S subunit, ATPase, 3,26S proteasome AAA-ATPase subunit RPT5, Proteasome 26S subunit ATPase 3, Proteasome subunit P50, TBP-126S protease regulatory subunit 6A, TBP1Tat-binding protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction