missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMC3 Monoclonal Antibody (1G10B7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548580
This item is not returnable.
View return policy
Description
PSMC3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA
This gene encodes a WASP interacting protein (WIP)-related protein. It has been shown that this protein has a role in the WASP-mediated organization of the actin cytoskeleton and that this protein is a potential link between the activated platelet-derived growth factor receptor and the actin polymerization machinery.
Specifications
| PSMC3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P17980, Q63569 | |
| PSMC3 | |
| Purified recombinant fragment of human PSMC3 (AA: 53-152) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Rat, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1G10B7 | |
| Unconjugated | |
| PSMC3 | |
| 26S protease regulatory subunit 6A; 26S proteasome AAA-ATPase subunit RPT5; 26S proteasome regulatory subunit 6A; human immunodeficiency virus tat transactivator binding protein-1; MGC8487; proteasome (prosome, macropain) 26S subunit, ATPase 3; proteasome (prosome, macropain) 26S subunit, ATPase, 3; proteasome 26S subunit ATPase 3; proteasome 26S subunit, ATPase 3; proteasome subunit P50; Psmc3; rpt5; Spermatogenic cell/sperm-associated Tat-binding protein homolog SATA; Tat binding protein 1; Tat-binding protein 1; TBP1; TBP-1; testicular secretory protein Li 42 | |
| Mouse | |
| Protein G | |
| RUO | |
| 29677, 5702 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction