missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19985
This item is not returnable.
View return policy
Description
PSMC6 Polyclonal specifically detects PSMC6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PSMC6 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| 26S protease regulatory subunit 10B, 26S protease regulatory subunit S10B, 26S proteasome AAA-ATPase subunit RPT4, CADP44, conserved ATPase domain protein 44, MGC12520, p42, P44, proteasome (prosome, macropain) 26S subunit, ATPase, 6, Proteasome 26S subunit ATPase 6, Proteasome subunit p42, SUG2 | |
| Rabbit | |
| 44 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.76mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P62333 | |
| PSMC6 | |
| Recombinant protein encompassing a sequence within the center region of human PSMC6. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5706 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction