missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD13 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99614AF350
This item is not returnable.
View return policy
Description
PSMD13 Polyclonal antibody specifically detects PSMD13 in Human samples. It is validated for Western Blot
Specifications
| PSMD13 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| HSPC027,26S proteasome regulatory subunit RPN9, p40.5,26S proteasome subunit p40.5,26S proteasome regulatory subunit p40.5, proteasome (prosome, macropain) 26S subunit, non-ATPase, 13,26S proteasome regulatory subunit S11, Rpn9,26S proteasome non-ATPase regulatory subunit 13, S11 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSMD13. | |
| 0.1 mL | |
| Cell Biology | |
| 5719 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction