missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | PSMD13 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PSMD13 Polyclonal antibody specifically detects PSMD13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| PSMD13 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 5719 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| HSPC027,26S proteasome regulatory subunit RPN9, p40.5,26S proteasome subunit p40.5,26S proteasome regulatory subunit p40.5, proteasome (prosome, macropain) 26S subunit, non-ATPase, 13,26S proteasome regulatory subunit S11, Rpn9,26S proteasome non-ATPase regulatory subunit 13, S11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human PSMD13 (NP_002808.3). MKDVPGFLQQSQNSGPGQPAVWHRLEELYTKKLWHQLTLQVLDFVQDPCFAQGDGLIKLYENFISEFEHRVNPLSLVEIILHVVRQMTDPNVALTFLEKTREKVKSSDEAVILCKTAIGALKLNIGDLQVTKETIEDVEEMLNNLPGVTSVHSRFYDLSSKYYQTIGNHASYYKDALRFLGCVDIKDLPVSEQQERAFTLGLAGLLGEGVFNFGELLMHPVLESLRNTDRQWLIDTLYAFNSGNVERFQT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title