missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMD13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558497
This item is not returnable.
View return policy
Description
Immunogen sequence: LEKTREKVKS SDEAVILCKT AIGALKLNIG DLQVTKETIE DVEEMLNNLP GVTSVHSRFY DLSSKYYQTI GNHASYYKDA LRFLGCVDIK DLPVSEQQE Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Acts as a regulatory subunit of the 26S proteasome which is involved in the ATP-dependent degradation of ubiquitinated proteins.
Specifications
| PSMD13 | |
| Polyclonal | |
| Unconjugated | |
| PSMD13 | |
| 26S proteasome non-ATPase regulatory subunit 13; 26S proteasome non-ATPase regulatory subunit 13 {ECO:0000250; 26S proteasome regulatory subunit p40.5; 26S proteasome regulatory subunit p40.5 {ECO:0000250; 26S proteasome regulatory subunit RPN9; 26S proteasome regulatory subunit S11; 26S proteasome regulatory subunit S11 {ECO:0000250; 26S proteasome subunit p40.5; EMBL:AAI58733.1, ECO:0000312; EMBL:AAI58733.1}; HSPC027; p40.5; proteasome (prosome, macropain) 26S subunit, non-ATPase, 13; proteasome 26S subunit, non-ATPase 13; PSMD13; psmd13 {ECO:0000312; RGD:1305236}; Rpn9; S11; UniProtKB:Q9UNM6, ECO:0000312; UniProtKB:Q9UNM6} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23997, 365388, 5719 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| B0BN93, Q9UNM6, Q9WVJ2 | |
| PSMD13 | |
| Recombinant protein corresponding to Human PSMD13. Recombinant protein control fragment (Product #RP-97532). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction