missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD14 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99615DL594
This item is not returnable.
View return policy
Description
PSMD14 Polyclonal antibody specifically detects PSMD14 in Human samples. It is validated for Western Blot
Specifications
| PSMD14 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| EC 3.1.2.15, EC 3.4.19.-, PAD1, POH126S proteasome non-ATPase regulatory subunit 14,26S proteasome regulatory subunit RPN11, proteasome (prosome, macropain) 26S subunit, non-ATPase, 14, Rpn11, RPN11,26S proteasome-associated PAD1 homolog 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSMD14. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10213 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction