missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD2 Monoclonal Antibody (OTI1D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503229S
This item is not returnable.
View return policy
Description
PSMD2 Monoclonal antibody specifically detects PSMD2 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| PSMD2 | |
| Monoclonal | |
| 0.73 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q03154 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 95 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI1D1 | |
| Unconjugated | |
| ACY1 | |
| 26S proteasome non-ATPase regulatory subunit 2; 26S proteasome non-ATPase regulatory subunit 2-like protein; 26S proteasome regulatory subunit RPN1; 26S proteasome regulatory subunit S2; 26S proteasome subunit p97; 55.11 protein; 9430095H01Rik; AA407121; F23F1.8; I79_017809; MGC14274; P97; proteasome (prosome, macropain) 26S subunit, non-ATPase, 2; proteasome 26S subunit, non-ATPase 2; protein 55.11; PSMD2; RPN1; S2; TEG-190; testis expressed gene 190; Tex190; TNFR-associated protein 2; TRAP2; Tumor necrosis factor type 1 receptor-associated protein 2 | |
| Full length human recombinant protein of human ACY1 produced in HEK293 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction