missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD2 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1G8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503270S
This item is not returnable.
View return policy
Description
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes one of the non-ATPase subunits of the 19S regulator lid. In addition to participation in proteasome function, this subunit may also participate in the TNF signalling pathway since it interacts with the tumor necrosis factor type 1 receptor. A pseudogene has been identified on chromosome 1.Specifications
| PSMD2 | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13200, Q4FZT9, Q8VDM4 | |
| Psmd2 | |
| Full length human recombit protein of human PSMD2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1G8 | |
| Unconjugated | |
| PSMD2 | |
| P97, RPN1, S2, TRAP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 21762, 287984, 478654, 5708 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction