missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSMD6 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5119363

 View more versions of this product

Product Code. 17790764

  • € 504.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PSMD6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a member of the protease subunit S10 family. The encoded protein is a subunit of the 26S proteasome which colocalizes with DNA damage foci and is involved in the ATP-dependent degradation of ubiquinated proteins. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PSMD6
Polyclonal
Unconjugated
PSMD6
2400006A19Rik; 26S proteasome non-ATPase regulatory subunit 6; 26S proteasome non-ATPase regulatory subunit 6-like protein; 26S proteasome non-ATPase regulatory subunit-like protein; 26S proteasome regulatory subunit RPN7; 26S proteasome regulatory subunit S10; breast cancer-associated protein SGA-113M; KIAA0107; p42A; P44s10; PFAAP4; Phosphonoformate immuno-associated protein 4; proteasome (prosome, macropain) 26S subunit, non-ATPase, 6; proteasome 26S subunit, non-ATPase 6; proteasome regulatory particle subunit p44S10; proteasome, 26S, non-ATPase regulatory subunit 6; PSMD6; Rpn7; S10; SGA-113M; vov; zgc:56471
Rabbit
Antigen affinity chromatography, Protein A
RUO
100519870, 9861
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q15008
PSMD6
Recombinant protein Proteasome 26S Subunit, Non ATPase 6. The antigen corresponds to amino acid range 1-389 of the target protein.
200 μL
Primary
Human, Pig
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.